Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 1 OD
SDP

Catalog No. p-7060000
Click to view available options
Quantity:
10 mL
100 mL
For Research Use Only
For Research Use Only
  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development

Specifications

Quantity 100 mL
Content And Storage Store at 4°C. Do not freeze.
Target 40nm Colloidal Gold Nanoparticles 1 OD
Product Type Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.