Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 20 OD
SDP

Catalog No. 2200010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-000-10 10 mL
22-001-00 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only
Catalog No. 22-000-10 Supplier Novus Biologicals™ Supplier No. 2200010
For Research Use Only

High efficiency antibody binding for maximum sensitivity

  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 10 mL
Target 40nm Colloidal Gold Nanoparticles 20 OD
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.