Learn More
Siemens Healthineers INNOVANCE™ D-Dimer Kit
Catalog No. p-7235902 Shop All Siemens Healthineers Products
Click to view available options
Quantity:
150 Tests/Kit
300 Tests/Kit
This item is not returnable. View return policy
Catalog No. 23-044-669
Supplier: Siemens Healthineers 10445981
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the quantitative determination of cross-linked fibrin degradation products (D-dimers) in human plasma on Siemens Sysmex™ Coagulation Systems.
Combining an excellent clinical sensitivity of >98.9% with a high negative predictive value (NPV) of 99%, the INNOVANCE™ D-Dimer assay's FDA-cleared exclusion claim gives clinicians greater confidence that translates into better patient management.
The fully automated INNOVANCE™ D-Dimer assay offers consistent performance on Siemens Healthineers and Sysmex coagulation instrument platforms for equivalent D-dimer test results. Speed and performance make INNOVANCE™ D-Dimer a robust, cost-effective assay for both routine and emergency use.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) D-Dimer Testing
Quantity 150 Tests/Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.