Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Acid-Base
GREENER_CHOICESCI_ED

Catalog No. S90731 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90731 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer.
Catalog No. S90731 Supplier Innovating Science™ Supplier No. IS8006
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer.

Includes

Materials for 15 groups: 25mL Hydrochloric Acid, 32% UN1789; 10g Sodium Hydroxide UN1823; 30mL Isopropyl Alcohol, 99% UN1219 (2); 30mL Acetone UN1090; teacher's guide; and student copy masters

In this experiment, determine the exact molarity of a base and an acid.

Standarize a solution of sodium hydroxide, approximately 0.01M, with a solid acid, potassium hydrogen phthalate, to determine the exact molarity of the base. The standardized base will be titrated against a monoprotic unknown acid to determine the molarity of the acid. A pH indicator and/or a pH meter will be used to determine the equivalence point.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine the exact molarity of a base and an acid
Includes 2 x 25ml 1m hydrochloric acid, 1 x 30ml 6m sodium hydroxide, 1 x 5g potassium hydrogen phthalate, 1 x 25ml 1% phenolphthalein, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 5.875 x 4.25 x 6.875 in.
Class Size 15 Lab Groups
Product Type Acid-Base Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.