Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Acid Base Indicators and Titrations
GREENER_CHOICESCI_ED

Catalog No. s97014 Shop All Aldon Products
Click to view available options
Grade:
7 to 14

Catalog No. S97014

Supplier: Innovating Science™ IS8027

Only null left
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 25mL Hydrochloric Acid, 32% UN1789; 10g Sodium Hydroxide UN1823; 30mL Isopropyl alcohol, 99% UN1219 (2); 30mL Acetone UN1090; teachers guide; and student copy masters

Examine acids and bases.

pH is the measure of the concentration of hydrogen ions in a solution. As this concentration can extend over several orders of magnitude, it is convenient to express it by means of logarithms of base ten. Distilled water has a pH of 7; so how is it possible that distilled water has free hydrogen ions?

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To examine acids and bases
Includes 1 x 25mL 32% hydrochloric acid, 1 x 10g sodium hydroxide, 2 x 30mL 99% isopropyl alcohol, 1 x 30mL acetone, Teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Acid Base Indicators and Titrations Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.