Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Activity Series
SCI_ED

Catalog No. s90727 Shop All Aldon Products
Click to view available options
:
AP Chemistry Kit: Activity Series
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Potassium Bromate and Strong inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Potassium Bromate and Strong inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.

Includes

Materials for 15 groups: Copper metal strips (15); Zinc metal pieces (60); Magnesium metal strips, 5 in. UN1869 (15); Lead metal strips 0.375 x 5 in. (1.0 x 12.7cm) (15); 50mL Cupric Nitrate, 0.1M solution (3); 50mL Zinc Nitrate, 0.1M solution (3); 50mL Magnesium Nitrate, 0.1M solution (3); 50mL Lead Nitrate, 0.1M solution (3); 4.3g Potassium Bromide; 1.2g Potassium Bromate UN1484; 100mL Sulfuric Acid, 0.5M solution; 100mL Chlorine Water solution UN1760; 100mL Iodine Water solution; 50mL Sodium Bromide, 0.1M solution (2); 50mL Potassium Iodide, 0.1N solution (2); 50mL Sodium Chloride, 0.1M solution (2); 100mL Mineral Oil (2); empty bottle labeled Bromine Water; teacher's guide; and student copy masters

In this experiment, empirically determine an activity series for a group of metals and a group of nonmetals.

An activity series of metals is a table of metals arranged in order of their decreasing chemical activity or the ease at which the metal will give up one or more electrons to form positive ions; this table is similar to the electrochemical series of elements. For example, if you take the group of metals magnesium, mercury and nickel, magnesium is the most reactive and mercury the least.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine an activity series for a group of metals and a group of nonmetals
Includes 45 Pieces Copper Metal, 45 Pieces Zinc Metal, 45 Pieces Magnesium Metal, 45 Pieces Lead Metal, 1 x 50mL 0.1M Copper Nitrate, 1 x 50mL 0.1M Zinc Nitrate, 1 x 50mL Magnesium Nitrate, 0.1M 1 x 50mL Lead Nitrate, 0.1M 1 x 2.15g Potassium Bromide, 1 x 0.6g Potassium Bromate, 1 x 25mL 1.0M Sulfuric Acid, 2 x 25mL Chlorine water, 2 x 25mL Iodine water, 2 x 25mL 0.1M Sodium Bromide, 2 x 25mL 0.1M Potassium Iodide, 2 x 25mL 0.1M Sodium Chloride, 2 x 75mL Mineral oil, 1 Empty bottle (for Bromine water), teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 7.25 x 7.25 x 8.375 in.
Class Size 15 Lab Groups
Product Type Activity Series Chemistry Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.