Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Electrochemical Cells
GREENER_CHOICESCI_ED

Catalog No. S90741 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S90741 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
Catalog No. S90741 Supplier Innovating Science™ Supplier No. IS8016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Includes

Materials for 15 groups: 500mL Cupric Sulfate, 0.5M solution (2); 25mL Bromothymol Blue, 0.04% solution; 500mL Sodium Sulfate, 1M solution (8); Copper metal strips (15); Magnesium metal strips 5 in. UN1869 (15); Dialysis tubing strips 6 in. (15); teacher's guide; and student copy masters

Electrolysis of water produces hydrogen and oxygen.

The tendency of oxidation-reduction reactions is to proceed to an equilibrium state. In electrochemical cells, these reactions provide another way for us to express the driving force in chemical reactions.
When reagents that accept or donate electrons are arranged so that the electrons can enter or leave the reaction through a metallic conductor, an electrochemical cell is established. A half-cell contains a metal in contact with a solution of its salt. Each metal will develop a different electrical potential based on its electron configuration.
The standard reduction potential listed in various references is the voltage that a half-cell develops when combined with a hydrogen half-cell. First, construct a simple chemical battery and determine from the standard reduction potentials what the output of the battery will be (if a voltmeter is available the actual and theoretical voltages can be compared). Second, construct an electrolysis cell and demonstrate how hydrogen and oxygen can be produced from the electrolysis of water.

  • Electrolysis cell is the reverse of a battery where a voltage higher than the reversible electromotive force of the cell is applied to the electrodes
  • In a cell containing water and an electrolyte, the oxidation reaction occurring at the anode produces oxygen gas while the reduction occurring at the cathode produces hydrogen gas
  • By the addition of a pH indicator, the oxidation-reduction reactions can be monitored visually
  • Gases produced are collected in inverted test tubes and the volumes of hydrogen and oxygen are compared

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To study electrolysis of water produces hydrogen and oxygen
Includes 2 x 500mL Cupric Sulfate, 0.5M solution, 1 x 25mL Bromothymol Blue, 0.04% solution, 8 x 500mL Sodium Sulfate, 1M solution, Copper Metal strips, Magnesium Metal strips 5 in., dialysis tubing strips 6 in., teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 10.25 x 10.25 x 8.5 in.
Class Size 15 Lab Groups
Product Type Electrochemical Cells Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.