Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Equilibrium Constant
SCI_ED

Catalog No. S90737 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90737 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90737 Supplier Innovating Science™ Supplier No. IS8012

missing translation for 'validMainframeMsg'

Includes

Materials for 15 groups: 500mL Iron(III) Nitrate, 0.20M solution; 100mL Potassium Thiocyanate, 0.002M solution (2); 100mL Nitric Acid, 0.5M solution UN2031; teacher's guide; and student copy masters

To determine the equilibrium constant for the formation of a colored species.

Set up a series of solutions with known concentrations of the FeSCN2+ ion and measure the absorbance of each solution spectrophotometrically. We will then construct a standard curve that can be used to determine the concentration of the unknown solutions of the next part.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine the equilibrium constant for the formation of a colored species
Includes 1 x 500ml iron (iii) nitrate, 0.2m, 2 x 100ml potassium thiocyanate, 0.002m 1 x 100ml nitric acid, 0.5m, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 8 x 8 x 8 in.
Class Size 15 Lab Groups
Product Type Ionic Compound K Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.