Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Ester Formation
GREENER_CHOICESCI_ED

Catalog No. S90730 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S90730 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone and Strong Inorganic Acid mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S90730 Supplier Innovating Science™ Supplier No. IS8005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone and Strong Inorganic Acid mists containing Sulfuric Acid, which are known to the State of California to cause cancer and reproductive harm.

Includes

Material for 15 groups: 25mL Acetic Acid, Glacial UN2789 (2); 15g Salicylic Acid; 25mL Octyl Alcohol (2); 25mL Methyl Alcohol UN1230 (2); 25mL Sulfuric Acid, ACS Concentrate UN1830 (2); teacher's guide; and student copy masters

Combines various alcohols with acetic and butyric acids to form esters, which are identifiable by their distinctive odors.

Carboxylic acids react with alcohols in the presence of a strong acid. This is an equilibrium reaction, which means both reactants and products are present at some concentration. For example, when 1 mole of acetic acid and 1 mole of ethyl alcohol are mixed, 0.667 mole of the ester and 0.667 of water are produced, and 0.333 mole of acid and 0.333 mole of alcohol remain at equilibrium.
The yield of ester can be increased either by removing one of the products of the reaction as it is formed or by increasing the concentration of one of the reactants. The normal procedure is to remove water using the dehydrating agent sulfuric acid, which also acts as the catalyst.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Combines various alcohols with acetic and butyric acids to form esters, which are identifiable by their distinctive odors
Includes 2 x 25ml acetic acid, 2 x 25ml n-butyric acid, 1 x 25ml n-butyl alcohol, 1 x 25ml octyl alcohol, 1 x 25ml methyl alcohol, 1 x 25ml ethyl alcohol, 1 x 25ml concentrated sulfuric acid, glass droppers, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 15 Lab Groups
Product Type Ester Formation Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.