Learn More
Innovating Science™ AP Chemistry Kits: Qualitative Analysis

Shop All Aldon Products


Description
Includes
Materials for 15 groups: 100mL Sodium Chloride, 0.1M solution; 100mL Sodium Bromide, 0.1M solution; 100mL Sodium Iodide, 0.1M solution; 100mL Sodium Nitrate, 0.1M solution; 100mL Sodium Phosphate, 0.1M solution; 100mL Sodium Sulfate, 0.1M solution; 25mL Silver Nitrate, 0.1N solution (2); 225mL Nitric Acid, 6M solution UN2031 (2); 100mL Ammonium Hydroxide, 6M solution UN2672 (5); 100mL Mineral Oil (2); 25mL Ferric Nitrate, 0.1M solution (2); 25mL Potassium Permanganate, 2% solution (2); 25mL Barium Chloride, 0.1N solution (2); 25mL Ammonium Molybdate, 0.1M solution (2); 25mL Diphenylamine Oxidation Reduction Indicator UN1830 (2); 25mL Stannous Chloride, 0.1M solution (2); teacher's guide; and student copy masters
Unknowns will be analyzed for one of six possible anions: Cl-, Br-, I-, PO42- NO3 and SO42-. A standard solution for each anion will also be run. Colorimetric or turbidimetric methods of analysis have been developed for most of the elements and ionic species. These methods can be employed to quickly determine the chemical composition of an unknown substance.
Specifications
Specifications
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | To determine the chemical composition of an unknown substance |
Includes | 1 x 100mL 0.1M Sodium Bromide, 1 x 100mL 0.1M Sodium Chloride, 1 x 100mL 0.1M Sodium Iodide, 1 x 100mL 0.1M Sodium Nitrate, 1 x 100mL 0.1M Sodium Phosphate,1 x 100mL Sodium Sulfate, 1 x 200mL Nitric Acid, 6M, 1 x 50mL Ammonium Hydroxide, 2 x 100mL Mineral oil, 1 x 75mL Ammonium Molybdate 0.1M, 1 x 75mL Diphenylamine reagent, 1 x 75mL Barium Chloride 0.1N, 1 x 100mL Potassium Permanganate 2%, 1 x 100mL Ferric Nitrate 0.1M, 1 x 50mL Silver Nitrate 0.1M, teacher's guide and student study guide copymasters |
Quantity | 1 |
Dimensions (L x W x H) | 12 x 12 x 12.25 in. |
Class Size | 15 Lab Groups |
Product Type | Qualitative Analysis Chemistry Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.