Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Stoichiometry
GREENER_CHOICESCI_ED

Catalog No. S90736 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90736 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S90736 Supplier Innovating Science™ Supplier No. IS8011
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

Material for 15 groups: 25 g Aluminum metal powder UN1396 (2); 250 mL Potassium Hydroxide, 3.0M solution UN1814 (2); 250 mL Sulfuric Acid, 3M solution UN2796 (4); 250 mL Ethanol/Water V/V 50/50% solution UN1170 (4); teacher's guide; and student copy masters

Performs a series of chemical reactions that lead to the synthesis of the ionic compound potassium alum.

Many combinations of mono and trivalent cations yield crystals of the same stoichiometry and structure. The crystals are normally in the form of an octahedron. Sodium, potassium and ammonium ions are often the monovalent species whereas aluminum and chromium are examples of the trivalent ions. Potassium alum, KAl(SO4)2 12H2O, is the most common and is used in water purification, paper manufacturing and as a mordant in dyeing. In this experiment we will perform a series of chemical reactions which lead to the synthesis of the ionic compound potassium alum. Kit Includes: 2 x 25 g Aluminum Metal Powder 2 x 250 mL Potassium Hydroxide, 3.0M Solution 4 x 250 mL Sulfuric Acid, 3M Solution 4 x 250 mL Ethanol/Water V/V 50/50% Solution

Specifications

Grade 9 to 12
For Use With (Application) Performs a series of chemical reactions that lead to the synthesis of the ionic compound potassium alum
Includes 2 x 25 g aluminum metal powder, 2 x 250 mL potassium hydroxide, 3.0m solution, 4 x 250 mL sulfuric acid, 3m solution, 4 x 250 mL ethanol/water v/v 50/50% solution, teacher's guide and student study guide copymasters
Class Size 15 Lab Groups
Product Type Stoichiometry Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.