Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ DNA Extraction Kit
GREENER_CHOICESCI_ED

Catalog No. s98659 Shop All Aldon Products
Click to view available options
Grade:
7 to 14
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Learn the history of the discovery of DNA and DNA structure.

  • Understand the nature of genetic inheritance and the role of DNA and proteins in genetic expression
  • Lab uses biological detergents, enzymes and ethanol to isolate DNA from plant material
  • Kit Includes:
    • 10% SDS/1.5% NaCl
    • Pepsin (to make 25 mL of 0.5% solution)
    • 95% ethanol (25 mL)
    • 20 zipper bags
    • 15 filters
    • 15 plastic tubes
    • 30 graduated pipets
    • 15 stirrers

Order Info

Enough materials for 15 groups.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Studying the structure of DNA along with the history of DNA discovery
Quantity 1 Kit
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type DNA Extraction Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.