Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Document Analysis: Comprehensive Lab Activities Educational Kit
SCI_ED

Catalog No. s28502 Shop All Aldon Products
Click to view available options
Class Size:
10 Lab Groups
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

10 x 20mL Ink Remover; 10 x 5mL Lemon Juice; 10 x 10mL Phenolphthalein; 10 x 5mL 1.0 M Sodium Hydroxide; 1 x 500mL Chromatography Solvent; 1 x 55mL Iodine Solution; 10 Pieces Chromatography Paper; 100 Pieces Paper Slips; 10 Pieces Paper A; 10 Pieces Paper B; 10 Pieces Paper C; 10 Pieces Paper D; 10 Pieces Paper E; 10 Pieces Paper F; 10 x 10 g Transfer Clay; 10 Forceps; 10 Magnifying glass; 50 Cotton Swabs; 2 Marker A; 2 Marker B; 2 Marker C; 2 Marker D; 2 #1 Pencil; 2 #2 Pencil; 2 #3 Pencil; 2 Yellow Highlighter; 10 Developing Chamber

Designed to introduce students to the field of document analysis. Innovating Science Document Analysis: Comprehensive Lab Activities Educational Kit, cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis.

  • Students make use of techniques like microscopy, chromatography and fuming to complete activities
  • Contains 10 unique sets of materials for 10 groups
  • Teacher’s Manual and Student Study Guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 10 Lab Groups
Product Type Document Analysis: Comprehensive Lab Activities Educational Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.