Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Dental Analysis Classroom Kit
GREENER_CHOICESCI_ED

Catalog No. S28526 Shop All Aldon Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28526 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28526 Supplier Aldon Corporation Supplier No. IS9021
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.

Forensic dentistry is defined as the branch of forensic medicine that deals with identification through teeth, dental apparatus and their markings. The activities in this kit will introduce students to the various techniques used in forensic dentistry while they try and identify the suspect in the case.

The eight different activities in this kit include the concepts of creating a dental chart, creating a dental casting, creating a bite impression, identification of a patient from dental records, metric analysis of castings and metric analysis of bite marks.

  • This kit includes enough materials for 30 students
  • Teacher Manual and Student Study guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Teaching the techniques used in dental analysis
Includes Includes materials for 8 activities x 30 students:
  • 1 lb pkg Dental Alginate
  • 2 Boxes Nitrile Glove, Medium
  • 12 Dental Impression Tray, Small
  • 2 x 900g Plaster of Paris
  • 24 Dental Impression Tray, Medium
  • 32 Plastic Cup, 3 oz
  • 15 Measuring Cup
  • 32 Plastic Cup, 7 oz
  • 15 Metric Ruler
  • 32 Plastic Scoop
  • 90 Tongue Depressor
  • 1 Box Toothpicks
  • 1 Box Wax Bite Plates
  • Teacher manual
  • Student Study Guide copymaster
Quantity 1
Class Size 30 Students
Product Type Forensic Dentistry Analysis Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.