Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Mendelian Genetics Kits
GREENER_CHOICESCI_ED

Catalog No. S05882 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05882 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05882 Supplier Innovating Science™ Supplier No. IS3010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this activity, students can simulate Mendel's work and determine patterns of inheritance.

When Austrian monk Gregor Mendel began observing and analyzing various traits of pea plants in the 1800s, little did he realize that his work would eventually become a foundation for the basis of modern genetics.

  • Using special chips and “double dice,” students can simulate both monohybrid and dihybrid crosses
  • After crosses, students determine genotypic and phenotypic ratios for select traits
  • Compare values to theoretical “ideal” values as put forth by Mendel
  • Contains enough materials for 15 groups

Kit Contents:
  • 15 monohybrid chips, female (red/yellow)
  • 15 monohybrid chips, male (red/white)
  • 15 dihybrid double dice, female (colored)
  • 15 dihybrid double dice, male (clear)
  • 15 plastic shaker cups

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Used to simulate mendelian genetics and determine patterns of inheritance
Includes 15 monohybrid chips, female (red/yellow), 15 monohybrid chips, male (red/white), 15 dihybrid double dice, female (colored), 15 dihybrid double dice, male (clear), 15 plastic shaker cups, teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Mendelian Genetics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.