Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction to Microbiology: Bacterial Growth and Staining Kit
GREENER_CHOICESCI_ED

Catalog No. S05984 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05984 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Catalog No. S05984 Supplier Innovating Science™ Supplier No. IS3014
Add to Cart
Add to Cart

WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Bacteria, good and bad, are all around us. In this activity students will collect and grow bacteria, then learn techniques for studying the bacteria they have grown. They will perform a simple staining technique on bacterial cells to study the morphology. They will also perform a differential staining technique, Gram staining, on bacterial cells and determine if the collected bacteria are Gram Positive, Gram- Negative, or both.

  • Students can perform a simple staining technique on bacterial cells to study the morphology of the bacteria with a microscope.
  • They will also perform a differential staining technique, Gram staining, on bacterial cells to determine if the collected bacteria are Gram-positive, Gram-negative, or both.
  • Kit provides enough materials for 15 setups.
  • Requires compound 4X, 10X or 40X microscope (not included).
  • Kit Includes:
    • Prepared nutrient agar (2 x 200 mL)
    • Methylene blue (30 mL)
    • Crystal violet (30 mL)
    • Gram's iodine (30 mL)
    • Safranin O (30 mL)
    • 95% Ethanol (4 x 30 mL)
    • 72 Microscope slides
    • 20 Sterile Petri dishes
    • 40 Sterile cotton swabs

Specifications

Grade 7 to 14
Description Educational Kit
Format Kit
Product Type Bacterial Growth and Staining Kit
Quantity 1 Kit
Color White
Languages English
For Use With (Application) Performing a simple staining technique on bacterial cells
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.