Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Introduction To Reaction Rates Educational Kit
GREENER_CHOICESCI_ED

Catalog No. s28484 Shop All Aldon Products
Click to view available options
Class Size:
10 Lab Groups
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol, and Acetyl Salicylic Acid/ Aspirin, which are known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol, and Acetyl Salicylic Acid/ Aspirin, which are known to the State of California to cause cancer and reproductive harm.

Designed to introduce the concepts of reaction rates through a series of experiments demonstrating the impact of concentration, heat, surface Area and catalysts. Innovating Science Introduction To Reaction Rates Educational Kit, helps students to understand various factors that influence the rate of a reaction.

  • Contains enough materials for 10 groups
  • Teacher's manual and Student Study Guide copymasters are included
  • Kit Includes:
    • 0.01M Sodium Hydroxide (500 mL)
    • Phenolphthalein Indicator (10 x 1 mL)
    • Iodine Clock Reagent A (300 mL)
    • Iodine Clock Reagent B (400 mL)
    • 0.15M Sodium Thiosulfate (1 L)
    • 2.0M Hydrochloric Acid (300 mL)
    • Copper Metal Powder (1 g)
    • 20 Effervescent Tablet
    • 20 Aspirin Tablets
    • 80 Reaction Vessels
    • 1 Pack Wooden Dowels

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1 Kit
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 10 Lab Groups
Product Type Introduction to Reaction Rates Educational Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.