Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Qualitative Coliform Test Kit
GREENER_CHOICESCI_ED

Catalog No. s06684 Shop All Aldon Products
Change view
Click to view available options
Class Size:
25 Students
Catalog No. Class Size
S06684 25 Students
1 options

Catalog No. S06684

Supplier: Innovating Science™ IS5031

Only null left
Add to Cart
Add to Cart

Includes

Instruction manual, 25 lactose tubes

Detect coliforms in water sample using a distinct color change test.

  • Coliforms are a broad group bacteria mostly found in the intestines of warm-blooded animals.
  • While these organisms are not typically associated with serious illness themselves, they are often used as indicator organisms.
  • The presence of coliforms may indicate a possible presence of other pathogenic or disease-causing organisms.
  • Easy-to-use and economical test kit, students simply add a small amount of collected water to a sample tube and incubate to get a distinct color change indicating that coliforms are present.
  • Reaction time: 24 to 48 hours incubation
  • Quantity: 25 tests

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes 25 Lactose Tubes
Quantity 1
Dimensions (L x W x H) 4 x 4 x 4 in.
Class Size 25 Students
Product Type Coliform Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.