Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Electrochemical Series
GREENER_CHOICESCI_ED

Catalog No. S40738 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40738 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40738 Supplier Innovating Science™ Supplier No. IS8030SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students will study electrochemical series.

  • The electrochemical series is built up by arranging various redox equilibria in order of their standard electrode potentials (redox potentials).
  • When a strip of metal (an electrode) is placed in water the metal has a tendency to go into solution as ions with a simultaneous build up of electrons on the metal strip.
  • This process produces an electrical potential difference between the metal and solution which is called an electrode potential (E°).
  • Kit contains enough material for 5 groups
  • Teacher's Manual and Student Study Guide copymasters are included

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) AP Chemistry
Includes 5 Copper Metal Strips; 5 Aluminum Metal Strips; 5 Magnesium Metal Strips; 5 Iron Metal Strips; 5 Zinc Metal Strips; 8 Filter Paper Strips; 1 ea EZ-Prep to make 500 mL of each of the following solutions: Copper Sulfate, 0.1M, Aluminum Sulfate, 0.1M, Magnesium Sulfate, 0.1M, Iron Sulfate, 0.1M, Zinc Sulfate, 0.1M
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6.5 x 2.8 in.
Class Size 5 Lab Groups
Product Type Chemistry Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.