Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Enzymes And Digestion
GREENER_CHOICESCI_ED

Catalog No. S40732 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40732 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. S40732 Supplier Innovating Science™ Supplier No. IS3009SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.

Students will expose three nutrients (carbohydrates, proteins, and lipids) to different digestive enzymes. The samples will be compared to nutrients to which no enzymes are added and chemical tests will be used to determine if the enzymes were effective in digesting the compounds.

  • Upon completion, students will not only understand the importance of the digestive system but also the vital role enzymes play in releasing nutrients from food and converting them to a form usable by the body.
  • Contains enough materials for 5 groups
  • Aligned to the Next Generation Science Standards (NGSS)
  • Disciplinary Core Ideas: LS1.A
  • Performance Expectations: MS-LS1-3
  • Cross Cutting Concepts: Systems and System Models; Science is a Human Endeavor
  • Engineering Practices: Engaging in Argument from Evidence

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Biology
Includes 1.75 g Pancreatin/Bile Salt; 1 g Albumin; 1 g Starch; 350mg Pepsin; 100 mg Amylase; 10 mL 0.1 m Sodium Hydroxide; 10 mL 0.1 mHydrochloric Acid; 10 mL Biuret Reagent; 10 mL Phenolphthalein 1% IPA; 10 mL IKI Dilute Lugols Solution; 10 mL Olive Oil; 4 Plastic Gaduated Pipettes
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 5 Lab Groups
Product Type Enzyme Activity Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.