Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Genetics Of Blood Types
GREENER_CHOICESCI_ED

Catalog No. S40735 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S40735 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40735 Supplier Innovating Science™ Supplier No. IS3102SGL
Add to Cart
Add to Cart

Blood type, an inherited characteristic, has use in everything from forensic investigations to medical procedures. In this activity, students will learn about the genetics that determine blood type and the possible inheritance patterns and how they express themselves.

  • Students will use simulated blood to determine the blood type of four unknown samples and use their results to assist in the resolution of a fictional paternity dispute.
  • Kit contains materials for 5 groups.
  • Aligned to the Next Generation Science Standards (NGSS).
  • Disciplinary Core Ideas: PS1.A; PS1.B; LS1.A; LS1.B; LS3.A; LS3.B
  • Performance Expectations: MS-PS1-2; MS-LS3-2; HS- LS3-1; HS- LS3-3
  • Cross Cutting Concepts: Structure and Function; Cause and Effect; Scale, Proportion and Quantity; Science is a Human Endeavor
  • Engineering Practices: Developing and Using Models; Asking Questions and Defining Problems; Analyzing and Interpreting Data

Specifications

Certifications/Compliance Next Gen Aligned
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Biology
Includes 4 Simulated Blood Samples; Mother; Child; Sample X; Sample Y; 1 btl Simulated Anti-A Serum; 1 btl Simulated Anti-B Serum; 20 Blood Typing Trays; 1 pkg Toothpicks
Material Cardboard Box Kit
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 5 Lab Groups
Product Type Genetics Concepts Kit
Show More Show Less

Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.