Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Innovating Science Testing Food For Nutrients
GREENER_CHOICESCI_ED

Catalog No. S05985 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05985 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05985 Supplier Innovating Science™ Supplier No. IS3013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helps students to understand the importance of proteins, carbohydrates, and lipids in living organisms.

  • Learns to identify a positive test result for proteins using biuret reagent
  • Examine the reaction between Benedict's reagent and a simple sugar
  • Iodine/potassium iodide helps in testing for the presence of starch and lipids using a fat-soluble dye

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Helps students to understand the importance of proteins, carbohydrates, and lipids in living organisms
Includes 25 mL iodine/potassium iodide solution, 25 mL biuret reagent, 25 mL benedict's solution, 25 mL 1% sudan iii stain, 25 mL vegetable oil, 10 g soluble starch, 10 g albumin, 10 g glucose
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Testing Food for Nutrients Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.