Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INO80 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP146157
Description
INO80 Polyclonal specifically detects INO80 in Human, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.Specifications
| INO80 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 3.6.1, EC 3.6.1.23, EC 3.6.4.12, hINO80INO80 complex subunit A, homolog of yeast INO80, Ino80, INO80 complex homolog 1 (S. cerevisiae), INO80 homolog (S. cerevisiae), INO80A, INOC1putative DNA helicase INO80 complex homolog 1, KIAA1259 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54617 | |
| Human, Rat, Bovine, Canine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:4000 | |
| Q9ULG1, Q6ZPV2 | |
| INO80 | |
| Peptide with sequence C-TQEDYDSNHFKAQ corresponding to internal region according to NP_060023.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction