Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INO80 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178758PCP
Description
INO80 Polyclonal antibody specifically detects INO80 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
INO80 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
EC 3.6.1, EC 3.6.1.23, EC 3.6.4.12, hINO80INO80 complex subunit A, homolog of yeast INO80, Ino80, INO80 complex homolog 1 (S. cerevisiae), INO80 homolog (S. cerevisiae), INO80A, INOC1putative DNA helicase INO80 complex homolog 1, KIAA1259 | |
The immunogen for this product maps to a region between residue 1400 and 1450 of human INO80 homolog using the numbering given in entry NP_060023.1 (GeneID 54617). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54617 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction