Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INO80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140070
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KRQQRKLNFL ITQTELYAHF MSRKRDMGHD GIQEEILRKL EDSSTQRQID IGGGVVVNIT QEDYDSNHFK AQALKNAENA YHIHQARTRS FDEDAKESR.
DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
Specifications
| INO80 | |
| Polyclonal | |
| Unconjugated | |
| Ino80 | |
| 2310079N15Rik; 4632409L19Rik; Chromatin-remodeling ATPase INO80; DNA helicase INO80; DNA helicase-related INO80 complex homolog 1; DNA helicase-related protein INO80; hINO80; homolog of yeast INO80; Ino80; INO80 complex homolog 1; INO80 complex subunit; INO80 complex subunit A; INO80 homolog; INO80 homolog (S. cerevisiae); INO80A; Inoc1; KIAA1259; putative DNA helicase INO80 complex homolog 1; RGD1310969; yeast INO80-like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULG1 | |
| Ino80 | |
| Recombinant protein corresponding to Human INO80. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction