Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INO80 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565296 Supplier Invitrogen™ Supplier No. PA565296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRQQRKLNFLI TQTELYAHFM SRKRDMGHDG IQEEILRKLE DSSTQRQIDI GGGVVVNITQ EDYDSNHFKA QALKNAENAY HIHQARTRSF DEDAKESR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INO80
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ino80
Gene Accession No. Q9ULG1
Gene Alias 2310079N15Rik; 4632409L19Rik; Chromatin-remodeling ATPase INO80; DNA helicase INO80; DNA helicase-related INO80 complex homolog 1; DNA helicase-related protein INO80; hINO80; homolog of yeast INO80; Ino80; INO80 complex homolog 1; INO80 complex subunit; INO80 complex subunit A; INO80 homolog; INO80 homolog (S. cerevisiae); INO80A; Inoc1; KIAA1259; putative DNA helicase INO80 complex homolog 1; RGD1310969; yeast INO80-like protein
Gene Symbols Ino80
Host Species Rabbit
Immunogen Recombinant Human INO80. Recombinant protein control fragment (Product #RP-106007).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.