Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INO80B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26890325UL
Description
INO80B Polyclonal antibody specifically detects INO80B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
INO80B | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
HMGIYL4, IES2, IES2 homolog, INO80 complex subunit B, PAP-1 binding protein, PAP-1BP, PAPA1, zinc finger, HIT type 4 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GQEEEEEQRWLDALEKGELDDNGDLKKEINERLLTARQRALLQKARSQPSPMLPLPVAEGCPPPALTEEMLLKREER | |
25 μL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
83444 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction