Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INO80B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570509 Supplier Invitrogen™ Supplier No. PA570509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

IN80B induces growth and cell cycle arrests at the G1 phase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INO80B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene INO80B
Gene Accession No. Q99PT3
Gene Alias 2510009I23Rik; hIes2; High mobility group AT-hook 1-like 4; HMG1YL4; Hmga1l4; HMGIYL4; IES2; IES2 homolog; INO80 complex subunit B; Ino80b; PAP (Pim-1 associated protein) associated protein 1; PAP-1 binding protein; PAP-1-associated protein 1; PAP-1BP; Papa1; PAPA-1; RGD1562258; zgc:153347; Zinc finger HIT domain-containing protein 4; zinc finger, HIT type 4; ZNHIT4
Gene Symbols INO80B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human INO80B (aa 209-258).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70020
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.