Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INO80B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584845 Supplier Invitrogen™ Supplier No. PA584845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQEEEEEQRW LDALEKGELD DNGDLKKEIN ERLLTARQRA LLQKARSQPS PMLPLPVAEG CPPPALTEEM LLKREER Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 96%.

IN80B induces growth and cell cycle arrests at the G1 phase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INO80B
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INO80B
Gene Accession No. Q9C086
Gene Alias 2510009I23Rik; hIes2; High mobility group AT-hook 1-like 4; HMG1YL4; Hmga1l4; HMGIYL4; IES2; IES2 homolog; INO80 complex subunit B; Ino80b; PAP (Pim-1 associated protein) associated protein 1; PAP-1 binding protein; PAP-1-associated protein 1; PAP-1BP; Papa1; PAPA-1; RGD1562258; zgc:153347; Zinc finger HIT domain-containing protein 4; zinc finger, HIT type 4; ZNHIT4
Gene Symbols INO80B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INO80B. Recombinant protein control fragment (Product #RP-108569).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.