Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INO80C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561869
Description
Immunogen sequence: GYGASKKKKA SASSFAQGIS MEAMSENKMV PSEFSTGPVE KAAKPLPFKD PNFVHSGHGG AVAGKKNRT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
INO80C is a proposed core component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair.
Specifications
INO80C | |
Polyclonal | |
Unconjugated | |
INO80C | |
C18orf37; hIes6; IES6; IES6 homolog; INO80 complex subunit C; INO80C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
125476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PI98 | |
INO80C | |
Recombinant protein corresponding to Human INO80C. Recombinant protein control fragment (Product #RP-98245). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction