Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INO80E Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298592AF700
Description
INO80E Polyclonal specifically detects INO80E in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
INO80E | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CCDC95, coiled-coil domain containing 95, Coiled-coil domain-containing protein 95, FLJ00079, FLJ90652, INO80 complex subunit E | |
Produced in rabbits immunized with E. coli-derived Human INO80E fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
283899 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction