Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INO80E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | INO80E |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
INO80E Polyclonal specifically detects INO80E in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
INO80E | |
Polyclonal | |
Rabbit | |
Human | |
CCDC95, coiled-coil domain containing 95, Coiled-coil domain-containing protein 95, FLJ00079, FLJ90652, INO80 complex subunit E | |
INO80E | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
283899 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LPRKLKMAVGPPDCPVGGPLTFPGRGSGAGVGTTLTPLPPPKMPPPTILSTVPRQMFSDAGSGDDALDGDDD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title