Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copan Diagnostics Inoculating Loops and Needles in Zip Lock Pack, Calibrated, Disposable

Click to view available options
Volume:
1 μL
10 μL
Length (Metric) Handle:
199 mm
201 mm
Color:
Light Blue
Light Green
Description
- Re-sealable bag allows the user to open and close the loop pack to remove individual loops
- Disposable, sterile plastic loops and needles manufactured from soft-flexible plastic
- Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
- Ultra smooth surfaces allow easy streaking without cutting or gouging agar surface
- Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
- Color coded for easy product identification
- Packed in tamper-proof bags to guarantee sterility
- Certificate of calibration included in each case
Specifications
Specifications
Disposable | Single-use |
Volume | 1 μL |
Material | Plastic |
Sterility | Sterile |
Color | Light Green |
Product Type | Inoculating Loop, Flexible |
Includes | Evans blue dye certificate of calibration |
Length (Metric) Handle | 199 mm |
Quantity | 1000 |
Packaging Type | Zip lock resealable bags |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction