Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle Braided/Twisted Wire Loop

Catalog No. 13820042 Shop All Fisherbrand Products
Click to view available options
Size:
2 mm
3 mm
4 mm
5 mm
Includes:
Twisted Nichrome wire with aluminum handle
Twisted Nichrome wire with insulated handle

Catalog No. 13-820-042

Only null left
Add to Cart
Add to Cart

Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.

Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.

  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Cools rapidly after sterilization
  • Loops Slide along agar surfaces without cutting or tearing material
  • Nichrome wire available in single or twisted configuration
  • Passivated
  • 203 mm (8 in.) handles, available with insulation
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Length (Metric) 280 mm
Length (English) 11 in.
Material Nichrome
Size 5 mm
Color Silver
Product Type Inoculating Loop
For Use With (Equipment) Petri-Dish etc
Gauge 24
Includes Twisted Nichrome wire with insulated handle
Autoclavable Autoclavable
For Use With (Application) Ideal for selecting, inoculating, or transferring microbiological samples
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.