Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Antibody (4G4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259471B
This item is not returnable.
View return policy
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Inorganic Pyrophosphatase/PPA1 | |
Monoclonal | |
Biotin | |
diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
Recombinant human Inorganic Pyrophosphatase/PPA1 (1-289aa) purified from E. coli | |
0.1 mL | |
metabolism, Signal Transduction | |
5464 | |
Store at 4°C in the dark. | |
IgG2b λ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
4G4 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction