Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Antibody (SR1552), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Inorganic Pyrophosphatase/PPA1 |
---|---|
Clone | SR1552 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Inorganic Pyrophosphatase/PPA1 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
A synthesized peptide derived from human Inorganic Pyrophosphatase/PPA1 (Uniprot #: Q15181) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1552 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
metabolism, Signal Transduction | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
5464 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title