Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
iNOS Antibody (4E5), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222119AF594
Description
iNOS Monoclonal specifically detects iNOS in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
iNOS | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
NOS2 | |
iNOS Antibody (4E5) was made to a purified recombinant fragment of human iNOS (C-terminus) expressed in E. coli. [UniProt# P35228] | |
Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission | |
4843 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
400000 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2 | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction