Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iNOS Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5067258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.04 mL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300605 200 μL
NB300605SS 0.04 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300605 Supplier Novus Biologicals Supplier No. NB300605
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 65 publications

iNOS Polyclonal specifically detects iNOS in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, In vitro assay.

Specifications

Antigen iNOS
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In vitro Assay
Classification Polyclonal
Concentration 0.7 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:800, Flow Cytometry reported in scientific literature (PMID 31536479), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen reported in scientific literature (PMID 35005642), Immunoblotting, In vitro assay reported in scientific literature (PMID 27998907)
Formulation Tris-Glycine with 0.05% Sodium Azide
Gene Accession No. P29477
Gene Alias EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
Gene Symbols NOS2
Host Species Rabbit
Immunogen iNOS Antibody was made to a synthetic peptide corresponding to residues D(17) L K E E K D I N N N V K K T(31) of mouse iNOS.
Molecular Weight of Antigen 131 kDa
Purification Method Affinity purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4843
Test Specificity This antibody detects iNOS. It does not detect other NOS isoforms.
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.