Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
iNOS Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299091V
Description
iNOS Polyclonal specifically detects iNOS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
iNOS | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human iNOS. | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission | |
4843 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction