Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
iNOS Antibody (SR1134), Novus Biologicals™

Rabbit Monoclonal Antibody
$313.06 - $438.32
Specifications
| Antigen | iNOS |
|---|---|
| Clone | SR1134 |
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
Description
iNOS Monoclonal antibody specifically detects iNOS in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| iNOS | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2 | |
| A synthesized peptide derived from human iNOS (Uniprot #: P35228) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1134 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4843 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title