Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iNOS Polyclonal Antibody, CoraLite594, Proteintech
SDP

Catalog No. 502257536 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7536 Supplier Proteintech Group Inc Supplier No. CL59418985100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

iNOS Polyclonal antibody specifically detects iNOS in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen iNOS
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate CoraLite 594
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PODXL
Gene Accession No. O00592
Gene Alias Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Form Liquid
Isotype IgG
Target Molecule Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.