Learn More
iNOS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 222261AP150UL
Description
The iNOS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human iNOS. This antibody recognizes human, mouse antigen. The iNOS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
iNOS | |
Polyclonal | |
Unconjugated | |
150 μL | |
4843 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
533 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.