Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonicotinic acid, 99%

Catalog No. AC122595000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC122595000

Supplier: Thermo Scientific Chemicals 122595000

Only null left
Add to Cart

CAS: 55-22-1 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-22-1
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key TWBYWOBDOCUKOW-UHFFFAOYSA-N
Synonym isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
PubChem CID 5922
ChEBI CHEBI:6032
IUPAC Name pyridine-4-carboxylic acid
SMILES C1=CN=CC=C1C(=O)O

Specifications

Melting Point 310°C to 315°C
Color Cream
Boiling Point 260°C (15.0 mmHg)
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 22,45
Merck Index 15,5233
Solubility Information Solubility in water: 5.2g/L (20°C). Other solubilities: practically insoluble in benzene,ether and,boiling alcohol
Formula Weight 123.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Isonicotinic acid
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 200-228-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.