Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Inositol hexaphosphoric acid, 50 wt% aqueous solution

Catalog No. p-148591
Click to view available options
Quantity:
250 mL
1 L
2.5 L

CAS: 83-86-3 | C6H18O24P6 | 660.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Inositol hexaphosphoric acid
Name Note 40-50 wt. % Aqueous Solution
CAS 83-86-3, 7732-18-5
Boiling Point 105.0°C
Molecular Formula C6H18O24P6
MDL Number MFCD00082309
Density 1.3700g/mL
Synonym phytic acid, phytate, fytic acid, inositol hexaphosphate, alkalovert, myo-inositol hexaphosphate, alkovert, phytine, acide fytique, acidum fyticum
Solubility Information Solubility in water: miscible.
InChI Key IMQLKJBTEOYOSI-GPIVLXJGSA-N
SMILES OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O
IUPAC Name (2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate
Molecular Weight (g/mol) 660.03
PubChem CID 890
Formula Weight 660.03
Packaging Glass bottle
Refractive Index 1.391
Specific Gravity 1.37
Quantity 250 mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.