Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP1 Monoclonal Antibody (1A6)
GREENER_CHOICE

Catalog No. PIMA519210
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519210 Supplier Invitrogen™ Supplier No. MA519210
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSDILRELLC VSEKAANIAR ACRQQEALFQ LLIEEKKEGE KNKKFAVDFK TLADVLVQEV IKQNMENKFP GLEKNIFGEE SNEFTNDWGE KITLRLCSTE EETAELLSKV LNGNKVASEA LARVVHQDVA FTDPTLDSTE INVPQDILGI WVDPIDSTYQ YIKGSADIKS NQGIFPCGLQ CVTILIGVYD IQTGVPLMGV INQPFVSRDP NTLRWKGQCY WGLSYMGTNM HSLQLTISRR NGSETHTGNT GSEAAFSPSF SAVISTSEKE TIKAALSRVC GDRIFGAAGA GYKSLCVVQG LVDIYIFSED TTFKWDSCAA HAILRAMGGG IVDLKECLER NPETGLDLPQ LVYHVENEGA AGVDRWANKG GLIAYRSRKR LETFLSLLVQ NLAPAETHT.

This gene encodes the enzyme inositol polyphosphate-1-phosphatase, one of the enzymes involved in phosphatidylinositol signaling pathways. This enzyme removes the phosphate group at position 1 of the inositol ring from the polyphosphates inositol 1,4-bisphosphate and inositol 1,3,4-trisphophosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A6
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene INPP1
Gene Accession No. P49441
Gene Alias 2300002C06Rik; AV137389; Inositol polyphosphate 1-phosphatase; inositol polyphosphate-1-phosphatase; Inpp1; IPP; IPPase
Gene Symbols INPP1
Host Species Mouse
Immunogen INPP1 (AAH15496, 1 a.a. ~ 399 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3628
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.