Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119206
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119206 200 μL
PIPA5121822 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119206 Supplier Invitrogen™ Supplier No. PA5119206
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

INPP4A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes an Mg++ independent enzyme that hydrolyzes the 4-position phosphate from the inositol ring of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4-trisphosphate, and inositol 3,4-bisphosphate. Multiple transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene INPP4A
Gene Accession No. Q96PE3
Gene Alias 107kDa; 4-Ptase-1; 9630012D15; D130048C09Rik; Inositol polyphosphate 4-phosphatase type I; Inositol polyphosphate 4-phosphatase-1; Inositol polyphosphate-4-phosphatase type I A; inositol polyphosphate-4-phosphatase, type 1; inositol polyphosphate-4-phosphatase, type I; inositol polyphosphate-4-phosphatase, type I, 107kD; inositol polyphosphate-4-phosphatase, type I, 107kDa; INPP4; Inpp4a; R74740; TVAS1; Type I inositol 3,4-bisphosphate 4-phosphatase; type I inositol-3; type I inositol-3,4-bisphosphate 4-phosphatase
Gene Symbols INPP4A
Host Species Rabbit
Immunogen Recombinant protein Inositol Polyphosphate-4-Phosphatase Type I 107kDa. The antigen corresponds to amino acid range 1-137 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.