Learn More
Invitrogen™ INPP4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596762
Description
Immunogen sequence: MTAREHSPRH GARARAMQRA STIDVAADML GLSLAGNIQD PDEPILEFSL ACSELHTPSL DRKPNSFVAV SVTTPPQAFW TKHAQTEIIE GTNNPIFLSS IAFFQDSLIN QMTQVKLSVY DVKDRSQGTM YLLGSGTFIV KDLLQDRHHR LHLTLRSAES DRVGNITVIG WQMEEKSDQR PPVTRSVDTV NGRMVLPVDE SLTEALGIRS KYASLRKDTL LKSVFGGAIC RMYRFPTTDG NHLRILEQMA; Positive Samples: Jurkat, MCF7, 293T, Mouse testis, Mouse skeletal muscle, Rat skeletal muscle.
This gene encodes an Mg++ independent enzyme that hydrolyzes the 4-position phosphate from the inositol ring of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4-trisphosphate, and inositol 3,4-bisphosphate. Multiple transcript variants encoding distinct isoforms have been described.
Specifications
INPP4A | |
Polyclonal | |
Unconjugated | |
INPP4A | |
107kDa; 4-Ptase-1; 9630012D15; D130048C09Rik; Inositol polyphosphate 4-phosphatase type I; Inositol polyphosphate 4-phosphatase-1; Inositol polyphosphate-4-phosphatase type I A; inositol polyphosphate-4-phosphatase, type 1; inositol polyphosphate-4-phosphatase, type I; inositol polyphosphate-4-phosphatase, type I, 107kD; inositol polyphosphate-4-phosphatase, type I, 107kDa; INPP4; Inpp4a; R74740; TVAS1; Type I inositol 3,4-bisphosphate 4-phosphatase; type I inositol-3; type I inositol-3,4-bisphosphate 4-phosphatase | |
Rabbit | |
Affinity chromatography | |
RUO | |
269180, 3631, 80849 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62784, Q96PE3, Q9EPW0 | |
INPP4A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human INPP4A (NP_0040181). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.