Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102936 Supplier Invitrogen™ Supplier No. PA5102936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total INPP4A.

This gene encodes an Mg++ independent enzyme that hydrolyzes the 4-position phosphate from the inositol ring of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4-trisphosphate, and inositol 3,4-bisphosphate. Multiple transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INPP4A
Gene Accession No. Q62784, Q96PE3, Q9EPW0
Gene Alias 107kDa; 4-Ptase-1; 9630012D15; D130048C09Rik; Inositol polyphosphate 4-phosphatase type I; Inositol polyphosphate 4-phosphatase-1; Inositol polyphosphate-4-phosphatase type I A; inositol polyphosphate-4-phosphatase, type 1; inositol polyphosphate-4-phosphatase, type I; inositol polyphosphate-4-phosphatase, type I, 107kD; inositol polyphosphate-4-phosphatase, type I, 107kDa; INPP4; Inpp4a; R74740; TVAS1; Type I inositol 3,4-bisphosphate 4-phosphatase; type I inositol-3; type I inositol-3,4-bisphosphate 4-phosphatase
Gene Symbols INPP4A
Host Species Rabbit
Immunogen A synthesized peptide derived from human INPP4A(Accession Q96PE3), corresponding to amino acid residues D348-C398.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269180, 3631, 80849
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.