Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INPP4B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282096
Description
INPP4B Polyclonal specifically detects INPP4B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.Specifications
INPP4B | |
Polyclonal | |
Unconjugated | |
PBS | |
INPP4B | |
INPP4B antibody was raised against a 17 amino acid peptide near the amino terminus of human INPP4B. The immunogen is located within the last 50 amino acids of INPP4B. Amino Acid Squence: QFTSIQKTPNEPQLEF | |
Affinity Purified | |
RUO | |
Primary | |
INPP4B antibody is human, mouse and rat reactive. At least two isoforms of INPP4B are known to exist; this antibody only detects the longer isoform. INPP4B antibody is predicted to not cross-react with INPP4A. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 2.5 ug/mL, Immunocytochemistry 2.5 ug/ml | |
inositol polyphosphate-4-phosphatase, type II, 105kDa, type II inositol-3,4-bisphosphate 4-phosphatase | |
Rabbit | |
100 kDa | |
0.1 mg | |
Cytokine Research | |
8821 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction