Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558057 Supplier Invitrogen™ Supplier No. PA558057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDSIPHHSDY DEEEWDRVWA NVGKSLNCII AMVDKLIERD GGSEGSGGNN DGEKEPSLTD AIPSHPREDW YEQLYPLILT LKDCMGEVVN RAKQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.

Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4- trisphosphate and inositol 1,4-bisphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP4B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INPP4B
Gene Accession No. O15327
Gene Alias D-myo-inositol-3,4-bisphosphate 4-phosphohydrolase; E130107I17Rik; inositol polyphosphate 4-phosphatase II; 4-phosphatase II; inositol polyphosphate 4-phosphatase type II; inositol polyphosphate 4-phosphatase type II beta isoform; inositol polyphosphate-4-phosphatase type II 105kD; inositol polyphosphate-4-phosphatase type II B; inositol polyphosphate-4-phosphatase, type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Inpp4b; phosphatidylinositol-3,4-bisphosphate 4-phosphatase; phosphoinositide 4-phosphatase; type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase
Gene Symbols INPP4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INPP4B. Recombinant protein control fragment (Product #RP-96621).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8821
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.